missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, Mikromol™

Product Code. 17973196
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17973196 Supplier Mikromol Supplier No. MM0877.130100
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butyrophenone
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material n-Butyrophenone
CAS 495-40-9
Analyte or Component Names n-Butyrophenone
Shipping Condition Room Temperature
Molecular Formula C10 H12 O
Certifications/Compliance ISO 9001
Synonym Butyrophenone, 1-Phenylbutan-1-one, n-Butyrophenone
InChI Formula InChI=1S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8H,2,6H2,1H3
SMILES CCCC(=O)c1ccccc1
IUPAC Name 1-phenylbutan-1-one
Molecular Weight (g/mol) 148.2
Quantity 100 mg
Formula Weight 148.0888 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.