missing translation for 'onlineSavingsMsg'
Learn More

Butyrophenone, 99%

Product Code. 11463764
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463764 250 g 250g
11408760 25 g 25g
11453764 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463764 Supplier Thermo Scientific Alfa Aesar Supplier No. A11345.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 495-40-9 | C10H12O | 148.205 g/mol

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-40-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009397
InChI Key FFSAXUULYPJSKH-UHFFFAOYSA-N
Synonym butyrophenone, n-butyrophenone, 1-butanone, 1-phenyl, phenyl propyl ketone, 1-phenyl-1-butanone, propyl phenyl ketone, unii-186i11wb5b, 1-phenyl-butan-1-one, propylphenylketone
PubChem CID 10315
IUPAC Name 1-phenylbutan-1-one
SMILES CCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 11°C to 13°C
Density 1.021
Boiling Point 222°C
Flash Point 88°C (190°F)
Odor Characteristic
Refractive Index 1.5195
Quantity 250 g
Beilstein 508305
Solubility Information Insoluble in water.
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material Butyrophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.