missing translation for 'onlineSavingsMsg'
Learn More

Butyronitrile, 99%

Product Code. 11377925
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377925 250 mL 250mL
11387925 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11377925 Supplier Thermo Scientific Alfa Aesar Supplier No. L02999.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-74-0 | C4H7N | 69.107 g/mol

Butyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.

Solubility
Miscible with benzene, alcohol, ether and dimethylformamide. Slightly soluble in water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 109-74-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Spezifikation

Melting Point -112°C
Density 0.792
Boiling Point 115°C to 117°C
Flash Point 18°C (64°F)
Linear Formula CH3CH2CH2C≡N
Refractive Index 1.384
Quantity 250 mL
UN Number UN2411
Beilstein 1361452
Merck Index 14,1597
Solubility Information Miscible with benzene,alcohol,ether and dimethylformamide. Slightly soluble in water.
Formula Weight 69.11
Percent Purity 99%
Chemical Name or Material Butyronitrile
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Butyronitrile
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Ergänzende Informationen
  • MIXTURE LIST-Contain: n-Butanenitrile

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.