missing translation for 'onlineSavingsMsg'
Learn More

Butyric acid hydrazide, 95%, Thermo Scientific™

Product Code. 15730809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730809 5 g 5g
15740809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15730809 Supplier Thermo Scientific Acros Supplier No. 147460050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3538-65-6
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
InChI Key FCCCRBDJBTVFSJ-UHFFFAOYSA-N
Synonym butyrohydrazide, butyric acid hydrazide, butanohydrazide, butyrylhydrazine, butanoic acid, hydrazide, n-butyric acid hydrazide, butyric hydrazide, butryic acid hydrazide, butanoic acid hydrazide, butyryl hydrazine
PubChem CID 77079
IUPAC Name butanehydrazide
SMILES CCCC(=O)NN

Specifications

Melting Point 43.0°C to 44.0°C
Boiling Point 120.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min
Linear Formula CH3CH2CH2C(O)NHNH2
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 102.14
Percent Purity 95%
Chemical Name or Material Butyric acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.