missing translation for 'onlineSavingsMsg'
Learn More

Butyraldoxime, TRC

Product Code. 30345607
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345607 Supplier Toronto Research Chemicals Supplier No. TRCB75545510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyraldoxime
CAS 110-69-0
Molecular Formula C4H9NO
Quantity 10 g
Synonym Butanaldoxime, Butylaldoxime, Butyraldoxime, NSC 1487, n-Butylaldoxime, n-Butyraldoxime
InChI Formula InChI=1S/C4H9NO/c1-2-3-4-5-6/h4,6H,2-3H2,1H3
SMILES CCCC=NO
IUPAC Name N-butylidenehydroxylamine
Molecular Weight (g/mol) 87.12
Formula Weight 87.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.