missing translation for 'onlineSavingsMsg'
Learn More

Butyraldoxime, 98%, Thermo Scientific™

Product Code. 11440263
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440263 100 g 100g
11450263 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440263 Supplier Thermo Scientific Alfa Aesar Supplier No. A11420.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-69-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00013943
InChI Key KGGVGTQEGGOZRN-SNAWJCMRSA-N
Synonym butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime
PubChem CID 5324278
IUPAC Name (NE)-N-butylidenehydroxylamine
SMILES CCCC=NO

Specifications

Melting Point -29°C
Density 0.91
Boiling Point 150°C to 152°C
Flash Point 58°C (136°F)
Linear Formula CH3CH2CH2CH=NOH
Refractive Index 1.437
Quantity 100 g
UN Number UN2840
Beilstein 1698799
Formula Weight 87.12
Percent Purity 98%
Chemical Name or Material Butyraldoxime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.