missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde, 98+%

Product Code. 11453777
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453777 2500 mL 2500mL
11443777 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453777 Supplier Thermo Scientific Alfa Aesar Supplier No. A18243.0F

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-72-8 | C4H8O | 72.11 g/mol

Butyraldehyde is used as an intermediate in the production of synthetic resins, rubber accelerators, tanning auxiliaries, perfumes, flavors, plasticizers and high molecular weight polymers. It acts as a chemical intermediate to prepare other chemicals such as 2-ethylhexanol, n-butanol, polyvinyl butyral (PVB), n-butyric acid and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyraldehyde is used as an intermediate in the production of synthetic resins, rubber accelerators, tanning auxiliaries, perfumes, flavors, plasticizers and high molecular weight polymers. It acts as a chemical intermediate to prepare other chemicals such as 2-ethylhexanol, n-butanol, polyvinyl butyral (PVB), n-butyric acid and pharmaceuticals.

Solubility
Miscible with water, ethanol, ether, ethyl acetate, acetone and toluene.

Notes
Air sensitive. Store in cool place. Incompatible with oxidizing agents, reducing agents, strong bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O

Specifications

Melting Point -99°C
Density 0.8
Boiling Point 75°C to 76°C
Flash Point −11°C (12°F)
Odor Pungent
Linear Formula CH3CH2CH2CHO
Refractive Index 1.38
Quantity 2500 mL
UN Number UN1129
Beilstein 506061
Sensitivity Air Sensitive
Merck Index 14,1591
Solubility Information Miscible with water,ethanol,ether,ethyl acetate,acetone and toluene.
Formula Weight 72.11
Percent Purity ≥98%
Chemical Name or Material Butyraldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.