missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyraldehyde 2,4-Dinitrophenylhydrazone, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Butyraldehyde-2,4-dinitrophenylhydrazone |
| CAS | 1527-98-6 |
| Molecular Formula | C10 H12 N4 O4 |
| Quantity | 1 g |
| Synonym | Butanal, (2,4-dinitrophenyl)hydrazone (9CI), Butyraldehyde, (2,4-dinitrophenyl)hydrazone (6CI,8CI), NSC 404127 |
| InChI Formula | InChI=1S/C10H12N4O4/c1-2-3-6-11-12-9-5-4-8(13(15)16)7-10(9)14(17)18/h4-7,12H,2-3H2,1H3 |
| SMILES | CCCC=NNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | N-(butylideneamino)-2,4-dinitroaniline |
| Molecular Weight (g/mol) | 252.23 |
| Formula Weight | 252.0859 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction