missing translation for 'onlineSavingsMsg'
Learn More

Butylparaben, MedChemExpress

Product Code. 16411118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411118 Supplier Medchem Express Supplier No. HYB1431
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Butylparaben is an organic compound, has proven to be a highly successful antimicrobial preservative in cosmetics, also used in medication suspensions, and as a flavoring additive in food.

Butylparaben is an organic compound, has proven to be a highly successful antimicrobial preservative in cosmetics, also used in medication suspensions, and as a flavoring additive in food.

Specifications

Chemical Name or Material Butylparaben
Physical Form Solid
CAS 94-26-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C11H14O3
For Use With (Application) COVID-19-immunoregulation
Synonym Butyl parahydroxybenzoate Butyl paraben Butyl 4-hydroxybenzoate
Solubility Information DMSO : ≥ 2.0 mg/mL (10.30 mM)
SMILES O=C(OCCCC)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 194.23
Formula Weight 194.23
Percent Purity 98.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.