missing translation for 'onlineSavingsMsg'
Learn More

Butylhydroxytoluene, LoGiCal

Product Code. 30460718
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30460718 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30460718

Brand: LoGiCal LGCFOR0679.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butylhydroxytoluene
CAS 128-37-0
Analyte or Component Names Butylhydroxytoluene
Shipping Condition Room Temperature
Molecular Formula C15 H24 O
Certifications/Compliance ISO 9001
Synonym Butylhydroxytoluene, 2,6-Bis(1,1-dimethylethyl)-4-methylphenol, 2,6-Bis(tert-butyl)-4-methylphenol, 2,6-Di(tert-butyl)hydroxytoluene, 2,6-Di-tert-butyl-4-cresol, 2,6-Di-tert-butyl-4-hydroxytoluene, 2,6-Di-tert-butyl-4-methyl-1-hydroxybenzene, 2,6-Di-tert-butyl-4-methylhydroxybenzene, 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresole, 2,6-Di-tert-butyl-p-methylphenol, 2,6-Di-tert-butylcresol, 2,6-Di-tert-butylmethylphenol, 2,6-tert-Butyl-4-methylphenol, 3,5-Di-tert-butyl-4-hydroxytoluene, 4-Hydroxy-3,5-di-tert-butyltoluene, 4-Methyl-2,6-bis(1,1-dimethylethyl)phenol, 4-Methyl-2,6-di-tert-butylphenol, AO 29, AO 4, AO 4K, AOX 4, AOX 4K, Advastab 401, Agidol, Agidol 1, Agidol 1A, Alkofen BP, Antage BHT, Antioxidant 264, Antioxidant 29, Antioxidant 30, Antioxidant 4, Antioxidant 4K, Antioxidant DBPC, Antioxidant KB, Antioxidant MPJ, Antioxidant T 501, Antox QT, B-NOX BHT-P, BAT, BHT, BHT 264, BHT Swanox, BHT-C, Buks, Butylated hydroxytoluene, CAO 1, CAO 3, Catalin CAO 3, Chemanox 11, DBPC, Dalpac, Deenax, Di-tert-Butyl-4-methylphenol, Di-tert-butyl-p-cresol, Di-tert-butylcresol, Dibunol, Dibutylated hydroxytoluene, Dibutylcresol, Dibutylhydroxytoluene, E 321, Impruvol, Ionol, Ionol 1, Ionol 330, Ionol BHT, Ionol CP, Ionol K, Ionole, Kerabit, Lowinox BHT, Lubrizol 817, NSC 6347, Naugard BHT, Nocrac 200, Nocrac 2000, Nonox TBC, P 21, P 21 (phenol), Parabar 441, Permanax BHT, Popol, Ralox BHT, Selosol H 633, Stavox, Sumilizer BHT, Sumilizer BHT-R, Sustane BHT, Swanox BHT, T 501, T 501 (phenol), Tenamene 3, Tenox BHT, Tonarol, Topanol, Topanol BHT, Topanol O, Topanol O-FG, Topanol OC, Topanol OL, Ultranox 226, Vanlube PC, Vanlube PCX, Vanox PC, Vianol, Vulkanox KB, Yoshinox BHT, o-Di-tert-butyl-p-methylphenol, BHT, 2,6-di-tert-butyl-4-methylphenol
InChI Formula InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3
Solution Type Neat
SMILES Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-methylphenol
Industry Type Clinical Testing
Molecular Weight (g/mol) 220.35
Quantity 10 mg
Formula Weight 220.1827 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.