missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylene Glycol (Butane-1,3-diol), Mikromol™
Description
Butylene Glycol (Butane-1,3-diol)
Specifications
Specifications
| Chemical Name or Material | 1,3-Butanediol |
| CAS | 107-88-0 |
| Analyte or Component Names | 1,3-Butanediol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H10 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1,3-Butanediol, (RS)-1,3-Butanediol, 1,3-Butylene glycol, 1,3-Dihydroxybutane, 1-Methyl-1,3-propanediol, 13BGK, 3-Hydroxy-1-butanol, Butylene glycol, DL-1,3-Butanediol, Jeechem Bugl, Methyltrimethylene glycol, NSC 402145, Niax DP 1022, (+/-)-Butane-1,3-diol, beta-Butylene glycol |
| InChI Formula | InChI=1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3 |
| SMILES | CC(O)CCO |
| IUPAC Name | butane-1,3-diol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?