missing translation for 'onlineSavingsMsg'
Learn More

Butyl Triacontanoate, TRC

Product Code. 30339480
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339480 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339480 Supplier Toronto Research Chemicals Supplier No. TRCB7501005MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyl Triacontanoate
CAS 105025-97-6
Molecular Formula C34 H68 O2
Quantity 5 mg
InChI Formula InChI=1S/C34H68O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31-32-34(35)36-33-6-4-2/h3-33H2,1-2H3
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OCCCC
IUPAC Name butyl triacontanoate
Molecular Weight (g/mol) 508.9
Formula Weight 508.5219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.