missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Butyl Sepharose™ High Performance Chromatography Media
Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
25mL
Description
- Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked 34μm agarose beads with aliphatic butyl functional groups
- High-resolution, capacity separations for recovery
- High ligand density for dynamic binding capacity even at low salt concentration
- Display minimum of ionic interactions
- Chemical stability for effective CIP and sanitization
- Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes
Specifications
Specifications
| Quantity | 200 mL |
| Stability | Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS |
| For Use With (Application) | For Intermediate and Polishing Step PurifiCatIon Steps |
| Storage Requirements | 4°C to 30°C |
| Binding Capacity | 38 mg b-lactoglobulin/mL mediμm |
| Product Type | Butyl Sepharose High Performance Media |
| Flow Rate | 100 to 200 cm/Hoμr |
| Particle Size | 24 to 44 μm |
| Max. Pressure | 300 kPa |
| Matrix | Cross-Linked Agarose |
| Show More |
Safety and Handling
Product Identifier
- Butyl Sepharose High Performance, 200 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction