missing translation for 'onlineSavingsMsg'
Learn More

Butyl Parahydroxybenzoate, Mikromol™

Product Code. 17911106
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17911106 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911106 Supplier Mikromol Supplier No. MM0496.00
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butyl Parahydroxybenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyl Parahydroxybenzoate
CAS 94-26-8
Analyte or Component Names Butyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C11 H14 O3
Certifications/Compliance ISO 17034
Synonym Butyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate, Ethyl Parahydroxybenzoate Imp. D (EP), Propyl Parahydroxybenzoate Imp. D (EP), Methyl Parahydroxybenzoate Imp. D (EP), 4-(Butoxycarbonyl)phenol, 4-Hydroxybenzoic acid butyl ester, Aseptoform Butyl, Butoben, Butyl 4-hydroxybenzoate, Butyl Butex, Butyl Tegosept, Butyl chemosept, Butyl p-hydroxybenzoate, Butyl parabens, Butyl parasept, Butylparaben, Mekkings B, Mekkins B, NSC 13164, NSC 8475, Nipabutyl, Preserval B, SPF, Solbrol B, Tegosept B, Tegosept Butyl, n-Butyl 4-hydroxybenzoate, n-Butyl p-hydroxybenzoate, n-Butylparaben, p-Hydroxybenzoic acid butyl ester, Ethyl Parahydroxybenzoate Impurity D, Methyl Parahydroxybenzoate Impurity D, Propyl Parahydroxybenzoate Impurity D
InChI Formula InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3
SMILES CCCCOC(=O)c1ccc(O)cc1
IUPAC Name butyl 4-hydroxybenzoate
Molecular Weight (g/mol) 194.23
Quantity 500 mg
Formula Weight 194.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.