missing translation for 'onlineSavingsMsg'
Learn More

Butyl p-toluenesulfonate, 99%

Product Code. 10055243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055243 500 g Glass bottle 500g
10481523 25 g Glass bottle 25g
10005982 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055243 Supplier Thermo Scientific Acros Supplier No. 380595000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 778-28-9 | C11H16O3S | 228.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-28-9
Molecular Formula C11H16O3S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00027203
InChI Key QYJXDIUNDMRLAO-UHFFFAOYSA-N
Synonym butyl p-toluenesulfonate, butyl tosylate, n-butyl p-toluenesulfonate, butyl p-methylbenzenesulfonate, butyl-p-toluenesulfonate, p-toluenesulfonic acid, butyl ester, benzenesulfonic acid, 4-methyl-, butyl ester, butyl toluene-4-sulphonate, p-toluenesulfonic acid n-butyl ester, p-toluene sulfonic acid, butyl ester
PubChem CID 13066
IUPAC Name butyl 4-methylbenzenesulfonate
SMILES CCCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Color Brown to Yellow
Density 1.12g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4SO3(CH2)3CH3
Refractive Index 1.5030 to 1.5050 (20°C, 589nm)
Quantity 500 g
Formula Weight 228.31
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Butyl p-toluenesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Butyl p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.