missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Oleate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Butyl Oleate |
| CAS | 142-77-8 |
| Molecular Formula | C22H42O2 |
| Quantity | 5 g |
| Synonym | (Z)-9-Octadecenoic Acid Butyl Ester, Oleic acid Butyl Ester, Advaplast 42, Butyl (Z)-9-Octadecenoate, Butyl cis-9-Octadecenoate, Hallco C 503, Kaolube 804, Kemester 4000, Kessco 554, Kesscoflex BO, Plasthall 914, Wilmar Butyl Oleate, Witcizer 100, Witcizer 101, n-Butyl Oleate |
| InChI Formula | InChI=1S/C22H42O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(23)24-21-6-4-2/h12-13H,3-11,14-21H2,1-2H3/b13-12- |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCCC |
| IUPAC Name | butyl (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 338.57 |
| Formula Weight | 338.3185 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction