missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Methanesulfonate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Butyl Methanesulfonate
Specifications
Specifications
| Chemical Name or Material | Butyl methanesulfonate |
| CAS | 1912-32-9 |
| Analyte or Component Names | Butyl methanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H12 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methanesulfonic acid butyl ester, Butyl mesylate, Butyl methanesulfonate, Methanesulfonic acid n-butyl ester, NSC 36060, n-Butyl methanesulfonate |
| InChI Formula | InChI=1S/C5H12O3S/c1-3-4-5-8-9(2,6)7/h3-5H2,1-2H3 |
| SMILES | CCCCOS(=O)(=O)C |
| IUPAC Name | butyl methanesulfonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction