missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methacrylate, TRC

Product Code. 30134472
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134472 50 mL 50mL
30005405 10 mL 10mL
17984795 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134472 Supplier Toronto Research Chemicals Supplier No. TRCB78949350ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-88-1
Molecular Formula C8 H14 O2
Molecular Weight (g/mol) 142.2
Synonym Methacrylic acid, butyl ester (6CI,8CI), 2-Methyl-2-propenoic acid butyl ester, Acryester B, Acryester BMA, Blemmer BMA, Butyl 2-methyl-2-propenoate, Butyl 2-methylacrylate, Butyl methacrylate, Light Ester NB, N 212, NSC 20956, n-Butyl methacrylate
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(=C)C

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3
Formula Weight 142.0994
Chemical Name or Material Methacrylic acid butyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.