missing translation for 'onlineSavingsMsg'
Learn More

Butyl butyrate, 98%, Thermo Scientific Chemicals

Product Code. 10697421
Click to view available options
Quantity:
5 mL
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-21-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009450
InChI Key XUPYJHCZDLZNFP-UHFFFAOYSA-N
Synonym butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
PubChem CID 7983
ChEBI CHEBI:87429
IUPAC Name butyl butanoate
SMILES CCCCOC(=O)CCC

Specifications

Acidity 1mg KOH/g max.
Melting Point -92.0°C
Color Colorless to Yellow
Density 0.8675g/mL
Boiling Point 164.0°C to 165.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CO2(CH2)3CH3
Refractive Index 1.4050 to 1.4070
Quantity 1 L
Beilstein 02, 271
Merck Index 15, 1558
Specific Gravity 0.8675
Solubility Information Solubility in water: 0.05%. Other solubilities: soluble in alcohol and ether
Formula Weight 144.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Butyl butyrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Butyl butyrate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'