missing translation for 'onlineSavingsMsg'
Learn More

Butyl benzoate, 98+%, Thermo Scientific Chemicals

Catalog Number 10319260
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
136-60-7
C11H14O2
178.23
MFCD00009439
XSIFPSYPOVKYCO-UHFFFAOYSA-N
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
8698
butyl benzoate
CCCCOC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
136-60-7
C11H14O2
178.23
MFCD00009439
XSIFPSYPOVKYCO-UHFFFAOYSA-N
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
8698
butyl benzoate
CCCCOC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -22°C
Color Yellow
Density 1.0000g/mL
Boiling Point 250°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COO(CH2)3CH3
Refractive Index 1.4960 to 1.498
Quantity 5 g
Merck Index 15, 1554
Specific Gravity 1
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 178.23
Percent Purity 98+%
Physical Form Oily Liquid
Chemical Name or Material Butyl benzoate, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'