missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, TRC

Product Code. 30339986
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339986 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339986 Supplier Toronto Research Chemicals Supplier No. TRCB692225100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyl Acetate
CAS 123-86-4
Molecular Formula C6 H12 O2
Quantity 100 mL
Synonym 1-Acetoxybutane, 1-Butyl acetate, Acetic acid n-butyl ester, Butyl acetate, Butyl ethanoate, NBA, NSC 9298, n-Butyl acetate
InChI Formula InChI=1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3
SMILES CCCCOC(=O)C
IUPAC Name butyl acetate
Molecular Weight (g/mol) 116.16
Formula Weight 116.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.