missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, Reagent Grade, 99.5%, Honeywell™

Product Code. 15655770 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655770 1 L Glass Bottle 1 L
15628420 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15655770 Supplier Honeywell Research Chemicals Supplier No. 5374541L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.88g/mL at 25°C
Boiling Point 126.2°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Refractive Index n20/D 1.394
Quantity 1 L
UN Number UN1123
Beilstein 1741921
Vapor Density 4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Percent Purity 99.50%
Chemical Name or Material Butyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.