missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™

Product Code. 15615600
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15615600

Brand: Honeywell-Riedel-de Haen 458602.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Density 0.88g/mL at 25°C
Boiling Point 126°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Refractive Index n20/D 1.394
Quantity 2.5 L
UN Number UN1123
Beilstein 1741921
Residue after Evaporation 0 to 0.001%
Vapor Density 4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material Butyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.