missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Methylbenzenesulfonate, TRC

Product Code. 30135799
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135799 5 g 5g
30006228 500 mg 500mg
30011186 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135799 Supplier Toronto Research Chemicals Supplier No. TRCB8104485G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-28-9
Molecular Formula C11H16O3S
Molecular Weight (g/mol) 228.31
Synonym Butyl 4-Toluenesulfonate, Butyl p-Methylbenzenesulfonate, Butyl p-Toluenesulfonate, Butyl p-Tosylate, NSC 6190, Toluene-4-sulfonic Acid Butyl Ester, n-Butyl p-Toluenesulfonate, n-Butyl Tosylate, 4-Methylbenzenesulfonic Acid Butyl Ester;
IUPAC Name butyl 4-methylbenzenesulfonate
SMILES CCCCOS(=O)(=O)c1ccc(C)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H16O3S/c1-3-4-9-14-15(12,13)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3
Formula Weight 228.082
Percent Purity >95%
Chemical Name or Material Butyl 4-Methylbenzenesulfonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.