missing translation for 'onlineSavingsMsg'
Learn More

Butoconazole Nitrate, Mikromol™

Product Code. 30049109
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049109 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049109 Supplier Mikromol Supplier No. MM1637.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butoconazole Nitrate
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Butoconazole Nitrate
CAS 64872-77-1
Analyte or Component Names Butoconazole Nitrate
Shipping Condition Room Temperature
Molecular Formula C19 H17 Cl3 N2 S . H N O3
Certifications/Compliance ISO 17034
Synonym (+/-)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, 1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, (+/-)-Butoconazole nitrate, Butaconazole nitrate, Butoconazole nitrate, Femstat, Gynomyk
InChI Formula InChI=1S/C19H17Cl3N2S.HNO3/c20-15-7-4-14(5-8-15)6-9-16(12-24-11-10-23-13-24)25-19-17(21)2-1-3-18(19)22;2-1(3)4/h1-5,7-8,10-11,13,16H,6,9,12H2;(H,2,3,4)
SMILES O[N+](=O)[O-].Clc1ccc(CCC(Cn2ccnc2)Sc3c(Cl)cccc3Cl)cc1
IUPAC Name 1-[4-(4-chlorophenyl)-2-(2,6-dichlorophenyl)sulfanylbutyl]imidazole;nitric acid
Molecular Weight (g/mol) 474.79
Quantity 250 mg
Formula Weight 473.0134 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.