missing translation for 'onlineSavingsMsg'
Learn More

Butoconazole nitrate, MedChemExpress

Product Code. 16461406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461406 200 mg 200mg
16408226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461406 Supplier Medchem Express Supplier No. HYB0293
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Butoconazole nitrate (RS 35887), an imidazole antifungal agent, is active against Candida spp. and effective against vaginal infections due to Candida albicans. Butoconazole nitrate is presumed to function as other imidazole derivatives via inhibition of steroid synthesis.

Butoconazole nitrate (RS 35887), an imidazole antifungal agent, is active against Candida spp. and effective against vaginal infections due to Candida albicans. Butoconazole nitrate is presumed to function as other imidazole derivatives via inhibition of steroid synthesis.

Specifications

Chemical Name or Material Butoconazole nitrate
Physical Form Solid
CAS 64872-77-1
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C19H18Cl3N3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym RS 35887
Solubility Information DMSO : ≥ 100 mg/mL (210.62 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=C(SC(CCC2=CC=C(Cl)C=C2)CN3C=CN=C3)C(Cl)=CC=C1.O[N+]([O-])=O
Molecular Weight (g/mol) 474.79
Formula Weight 474.79
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.