missing translation for 'onlineSavingsMsg'
Learn More

Butenafine Hydrochloride, TRC

Product Code. 30332595
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332595 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332595 Supplier Toronto Research Chemicals Supplier No. TRCB690195250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butenafine Hydrochloride
CAS 101827-46-7
Purity Grade Notes HPLC
Molecular Formula C23 H27 N . Cl H
Quantity 250 mg
Synonym 1-Naphthalenemethanamine, N-[[4-(1,1-dimethylethyl)phenyl]methyl]-N-methyl-, hydrochloride (1:1), 1-Naphthalenemethanamine, N-[[4-(1,1-dimethylethyl)phenyl]methyl]-N-methyl-, hydrochloride (9CI), Butenafine hydrochloride, KP 363, Mentax, Scorba
InChI Formula InChI=1S/C23H27N.ClH/c1-23(2,3)21-14-12-18(13-15-21)16-24(4)17-20-10-7-9-19-8-5-6-11-22(19)20;/h5-15H,16-17H2,1-4H3;1H
SMILES Cl.CN(Cc1ccc(cc1)C(C)(C)C)Cc2cccc3ccccc23
IUPAC Name 1-(4-tert-butylphenyl)-N-methyl-N-(naphthalen-1-ylmethyl)methanamine;hydrochloride
Molecular Weight (g/mol) 353.93
Formula Weight 353.191
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.