missing translation for 'onlineSavingsMsg'
Learn More

Butenafine Hydrochloride, MedChemExpress

Product Code. 16451846
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451846 500 mg 500mg
16481669 1 g 1g
16449827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451846 Supplier Medchem Express Supplier No. HY17396
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.

Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.

Specifications

Chemical Name or Material Butenafine Hydrochloride
Physical Form Solid
CAS 101827-46-7
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C23H28ClN
For Use With (Application) COVID-19-immunoregulation
Synonym KP363 Hydrochloride
Solubility Information DMSO : 25 mg/mL (70.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.83 mM; Need ultrasonic)
SMILES CN(CC1=CC=C(C(C)(C)C)C=C1)CC2=C3C=CC=CC3=CC=C2.[H]Cl
Molecular Weight (g/mol) 353.93
Formula Weight 353.93
Percent Purity 99.96%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.