missing translation for 'onlineSavingsMsg'
Learn More

Butein, MedChemExpress

Product Code. 16437078
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437078

Brand: Medchem Express HY16558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Butein is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC).

Butein is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC).

Specifications

Chemical Name or Material Butein
CAS 487-52-5
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C15H12O5
Quantity 10 mg
Synonym 2’,3,4,4’-tetrahydroxy Chalcone
Solubility Information DMSO : 50 mg/mL (183.65 mM; Need ultrasonic)
SMILES OC1=CC=C(C(/C=C/C2=CC(O)=C(O)C=C2)=O)C(O)=C1
Molecular Weight (g/mol) 272.25
Formula Weight 272.25
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.