missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butein, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Description
Butein is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC).
Specifications
Specifications
| Chemical Name or Material | Butein |
| CAS | 487-52-5 |
| Color | Brownish Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H12O5 |
| Quantity | 5 mg |
| Synonym | 2’,3,4,4’-tetrahydroxy Chalcone |
| Solubility Information | DMSO : 50 mg/mL (183.65 mM; Need ultrasonic) |
| SMILES | OC1=CC=C(C(/C=C/C2=CC(O)=C(O)C=C2)=O)C(O)=C1 |
| Molecular Weight (g/mol) | 272.25 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction