missing translation for 'onlineSavingsMsg'
Learn More

Butane-1,4-disulfonic Acid Disodium Salt, TRC

Product Code. 17904245
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904245 25 g 25g
17940365 1 g 1g
17923435 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904245 Supplier Toronto Research Chemicals Supplier No. TRCB68988025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36589-61-4
Molecular Formula C4 H8 O6 S2 . 2 Na
Molecular Weight (g/mol) 262.21
Synonym 1,4-Butanedisulfonic acid, sodium salt (1:2), 1,4-Butanedisulfonic acid, disodium salt (9CI), Disodium butane-1,4-disulfonate, 1,4-Butanedisulfonic acid disodium salt, 1,4-Disulfobutane disodium salt, Butane-1,4-disulfonic acid disodium salt
IUPAC Name disodium;butane-1,4-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)CCCCS(=O)(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H10O6S2.2Na/c5-11(6,7)3-1-2-4-12(8,9)10;;/h1-4H2,(H,5,6,7)(H,8,9,10);;/q;2*+1/p-2
Formula Weight 261.9558
Percent Purity >95%
Chemical Name or Material Butane-1,4-disulfonic Acid Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.