missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butamirate Citrate, Mikromol™
Description
Butamirate Citrate
Specifications
Specifications
| Chemical Name or Material | Butamirate Citrate |
| CAS | 18109-81-4 |
| Analyte or Component Names | Butamirate Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H29 N O3 . C6 H8 O7 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzeneacetic acid, alpha-ethyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Butyric acid, 2-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, citrate (1:1) (8CI), Ethanol, 2-[2-(diethylamino)ethoxy]-, 2-phenylbutyrate (ester), citrate (1:1) (8CI), 2-[2-(Diethylamino)ethoxy]ethyl-2-phenylbutyrate citrate (1:1), Abbott 36581, Acodeen, Butamirate citrate, Cough Cut, HH 197, Panatus, Sincodeen, Sincodin, Sincodix, Sinecod, Sinecond, Oxeladin impurity D, 2-(2-diethylaminoethoxy)ethyl 2-phenyl-butyrate dihydrogen citrate, Oxeladin Hydrogen Citrate Impurity D as Citrate, Oxeladin Impurity D as Citrate |
| InChI Formula | InChI=1S/C18H29NO3.C6H8O7/c1-4-17(16-10-8-7-9-11-16)18(20)22-15-14-21-13-12-19(5-2)6-3;7-3(8)1-6(13,5(11)12)2-4(9)10/h7-11,17H,4-6,12-15H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CCC(C(=O)OCCOCCN(CC)CC)c1ccccc1.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 2-[2-(diethylamino)ethoxy]ethyl 2-phenylbutanoate;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?