missing translation for 'onlineSavingsMsg'
Learn More

Butafosfan, TRC

Product Code. 30009821
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009821 1 g 1g
17951905 100 mg 100mg
17942355 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009821 Supplier Toronto Research Chemicals Supplier No. TRCB6894101G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17316-67-5
Molecular Formula C7 H18 N O2 P
Molecular Weight (g/mol) 179.2
Synonym Butafosfan, Butaphosphan, [1-(Butylamino)-1-methylethyl]phosphinic acid, 1-(Butylamino)-1-methylethylphosphonous acid, P-[1-(Butylamino)-1-methylethyl]phosphinic acid
IUPAC Name 2-(butylamino)propan-2-ylphosphinic acid
SMILES CCCCNC(C)(C)P(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H18NO2P/c1-4-5-6-8-7(2,3)11(9)10/h8,11H,4-6H2,1-3H3,(H,9,10)
Formula Weight 179.1075
Percent Purity >95%
Chemical Name or Material Butafosfan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.