missing translation for 'onlineSavingsMsg'
Learn More

Butadiene diepoxide, 95%, Thermo Scientific™

Product Code. 11346716
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346716 5 g 5g
11336716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346716 Supplier Thermo Scientific Alfa Aesar Supplier No. L03919.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Butadiene diepoxide is used as a research chemical, as a chemical intermediate, as a curing agent for polymers, as a cross-linking agent for textile fabrics, and to prevent microbial spoilage. It is also been used to synthesize erythritol and pharmaceuticals.

Solubility
Soluble in water (1000 g/L).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1464-53-5
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005120
InChI Key ZFIVKAOQEXOYFY-UHFFFAOYSA-N
Synonym diepoxybutane, 1,3-butadiene diepoxide, 2,2'-bioxirane, butadiene diepoxide, dioxybutadiene, butadiene dioxide, bioxirane, butane diepoxide, 1,2:3,4-diepoxybutane, butadiendioxyd
PubChem CID 11254
ChEBI CHEBI:23704
IUPAC Name 2-(oxiran-2-yl)oxirane
SMILES C1C(O1)C2CO2

Specifications

Melting Point -19°C
Density 1.112
Boiling Point 56°C to 58°C (25 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.434
Quantity 5 g
UN Number UN2929
Beilstein 79831
Merck Index 14,3676
Solubility Information Soluble in water (1000g/L).
Formula Weight 86.09
Percent Purity 95%
Chemical Name or Material Butadiene diepoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.