missing translation for 'onlineSavingsMsg'
Learn More

Butachlor, Dr. Ehrenstorfer

Product Code. 30309217
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30309217

Brand: Dr. Ehrenstorfer DREC10860000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butachlor
CAS 23184-66-9
Analyte or Component Names Butachlor
Shipping Condition Room Temperature
Molecular Formula C17 H26 Cl N O2
Certifications/Compliance ISO 17034
Synonym Acetanilide, N-(butoxymethyl)-2-chloro-2',6'-diethyl- (8CI), N-(Butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetanilide, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetoanilide, 2',6'-Diethyl-N-butoxymethyl-2-chloroacetanilide, 2',6'-Diethyl-N-butoxymethyl-α-chloroacetanilide, Bilchlor, Butaclor, CP 53619, Delchlor, Delchlor 5G, Dingcaoan, Hiltachlor, Machet, Machete, Machete (herbicide), Machette, N-(Butoxymethyl)-2-chloro-2',6'-diethylacetanilide, NSC 221683
InChI Formula InChI=1S/C17H26ClNO2/c1-4-7-11-21-13-19(16(20)12-18)17-14(5-2)9-8-10-15(17)6-3/h8-10H,4-7,11-13H2,1-3H3
Solution Type Neat
SMILES CCCCOCN(C(=O)CCl)c1c(CC)cccc1CC
IUPAC Name N-(butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide
Industry Type Environmental Testing
Molecular Weight (g/mol) 311.85
Quantity 100 mg
Formula Weight 311.1652 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.