missing translation for 'onlineSavingsMsg'
Learn More

But-1-ene (10% in Hexanes), TRC

Product Code. 30134889
Change view
Click to view available options
Quantity:
1 mL
2 mL
10 mL
Unit Size:
10mL
1mL
2mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134889 2 mL 2mL
30134556 1 mL 1mL
30010869 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134889 Supplier Toronto Research Chemicals Supplier No. TRCB7544532ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-98-9
Molecular Formula C4 H8
Molecular Weight (g/mol) 56.11
Synonym 1-Butene, 1-Butylene, Butene-1, Ethylethylene, α-Butene, α-Butylene
IUPAC Name but-1-ene
SMILES CCC=C

Specifications

Quantity 2 mL
InChI Formula InChI=1S/C4H8/c1-3-4-2/h3H,1,4H2,2H3
Formula Weight 56.0626
Concentration or Composition (by Analyte or Components) 1-Butene 10%
Chemical Name or Material 1-Butene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.