missing translation for 'onlineSavingsMsg'
Learn More

Buspirone-d8 HCl (n-butyl-d8), TRC

Product Code. 30337292
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337292 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30337292 Supplier Toronto Research Chemicals Supplier No. TRCB6898511MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buspirone-d8 HCl (n-butyl-d8)
CAS 1216761-39-5
Purity Grade Notes HPLC
Molecular Formula C21 D8 H23 N5 O2 . Cl H
Quantity 1 mg
Synonym 8-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl-1,1,2,2,3,3,4,4-d8]-8-azaspiro[4.5]decane-7,9-dione hydrochloride, Buspirone-d8 hydrochloride, Buspirone-D8 (1,4-diaminobutyl-D8) hydrochloride, Buspirone-d8 HCl (n-butyl-d8)
InChI Formula InChI=1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H/i3D2,4D2,10D2,11D2;
SMILES Cl.[2H]C([2H])(N1CCN(CC1)c2ncccn2)C([2H])([2H])C([2H])([2H])C([2H])([2H])N3C(=O)CC4(CCCC4)CC3=O
IUPAC Name 8-[1,1,2,2,3,3,4,4-octadeuterio-4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
Molecular Weight (g/mol) 430.0134
Formula Weight 429.2747
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.