missing translation for 'onlineSavingsMsg'
Learn More

Jaytec Glass™ Burette, Class B, Two Way Stopcock with PTFE Key

Product Code. 15775862
Change view
Click to view available options
Type: Two way burette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15775862 Type: Two way burette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15775862 Supplier Jaytec Glass™ Supplier No. WJ.278

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Two Way Borosilicate Glass Burette, with interchangeable PTFE Key and permanent enamel graduations.

  • A two way stopcock has an extra side arm for adding solution to the burette from a reservoir, avoiding the need to fill from the top with a funnel
  • Useful in situations where a large number of titrations are required or hazardous solutions are being used
  • Manufactured in accordance with BS EN ISO 385:2005

Specifications

Autoclavable Autoclavable
Color Clear
Capacity (Metric) 50 mL
For Use With (Equipment) Laboratory Stand, Burette Clamp
Material Borosilicate Glass
Includes PTFE key
Plug Material PTFE
Resolution 0.1 mL
Scale Permanent enamel graduations
Stopcock Two way
Stopcock Material Glass
Tolerance ±0.1 mL
Type Two-way Buret
Class Class B
Certifications/Compliance DIN EN ISO 385
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.