Learn More
S Murray™ Burette Clamps
Click to view available options
Unit Size:
Pack of 10
Description
- One-piece design molded in Delrin
- Suitable for 50mL burettes
- Without rod
- Supplied in packs of 10
Order Info
dimForUseWithEquipment
Specifications
Specifications
| No. per Pack | 10 |
| Type | Buret Clamp |