missing translation for 'onlineSavingsMsg'
Learn More

Bupropion Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463778
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463778 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463778 Supplier LoGiCal Supplier No. LGCAMP0745.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bupropion (hydrochloride)
CAS 31677-93-7
Analyte or Component Names Bupropion (hydrochloride)
Shipping Condition Ice pack (-20°C)
Molecular Formula C13 H18 Cl N O . Cl H
Certifications/Compliance ISO 17034
Synonym 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1), 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (9CI), Bupropion hydrochloride, NSC 315851, Wellbatrin, Wellbutrin, Zyban, Zyban (pharmaceutical), 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride, (+/-)-, Propiophenone, 2-(tert-butylamino)-3'-chloro-, hydrochloride, (+/-)- (8CI), DL-alpha-t-Butylamino-3-chloropropiophenone hydrochloride, m-Chloro-alpha-tert-butylaminopropiophenone hydrochloride, alpha-(tert-Butylamino)-m-chloropropiophenone hydrochloride, 3134W92, 269W93, GI 267196X, 269W93, 4U65 HCl
InChI Formula InChI=1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H
Solution Type Single Solution
SMILES Cl.CC(NC(C)(C)C)C(=O)c1cccc(Cl)c1
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 276.2
Quantity 1 mL
Formula Weight 275.0844 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.