missing translation for 'onlineSavingsMsg'
Learn More

Bupropion.HCl, Lipomed

Product Code. 30459979
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459979 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459979 Supplier Lipomed Supplier No. LPMBPR1544HC50
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Bupropion.HCl
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Bupropion (hydrochloride)
CAS 31677-93-7
Analyte or Component Names Bupropion (hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C13 H18 Cl N O . Cl H
Synonym 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1), 1-Propanone, 1-(3-chlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (9 CI), Bupropion hydrochloride, NSC 315851, Wellbatrin, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X, GI 267196 X
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CC(NC(C)(C)C)C(=O)c1cccc(Cl)c1
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 276.2
Quantity 50 mg
Formula Weight 275.0844 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.