missing translation for 'onlineSavingsMsg'
Learn More

Bupivacaine Hydrochloride Monohydrate, Mikromol™

Product Code. 18001170
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18001170 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001170 Supplier Mikromol Supplier No. MM0613.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bupivacaine Hydrochloride Monohydrate
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Bupivacaine Hydrochloride Monohydrate
CAS 73360-54-0
Analyte or Component Names Bupivacaine Hydrochloride Monohydrate
Shipping Condition Room Temperature
Molecular Formula C18 H28 N2 O . Cl H . H2 O
Certifications/Compliance ISO 17034
Synonym Bupivacaine Hydrochloride Monohydrate, 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide monohydrochloride monohydrate
InChI Formula InChI=1S/C18H28N2O.ClH.H2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H;1H2
SMILES O.Cl.CCCCN1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC Name 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride
Molecular Weight (g/mol) 342.9
Quantity 250 mg
Formula Weight 342.2074 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.