missing translation for 'onlineSavingsMsg'
Learn More

Bupivacaine hydrochloride monohydrate, 98+%

Product Code. 15475139
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475139 25 g 25g
15455139 1 g 1g
15465139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475139 Supplier Thermo Scientific Alfa Aesar Supplier No. J62835.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits TREK-1 channels and depolarizes the cell membrane | CAS: 73360-54-0 | C18H31ClN2O2 | 342.908 g/mol

It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

Solubility
Soluble in water to 50 mM, in DMSO to 100 mM and in ethanol to 100 mM.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73360-54-0
Molecular Formula C18H31ClN2O2
Molecular Weight (g/mol) 342.908
MDL Number MFCD00078956
InChI Key HUCIWBPMHXGLFM-UHFFFAOYSA-N
Synonym bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate
PubChem CID 5282419
IUPAC Name 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride
SMILES CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl

Specifications

Melting Point 255°C to 259°C
Color White
Quantity 25 g
UN Number UN2811
Merck Index 14,1495
Solubility Information Soluble in water to 50 mM,in DMSO to 100 mM and in ethanol to 100 mM.
Formula Weight 342.9
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Bupivacaine hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.