missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buparvaquone, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Buparvaquone is a hydroxynaphthoquinone antiprotozoal drug related to parvaquone and atovaquone.
Specifications
Specifications
| Chemical Name or Material | Buparvaquone |
| CAS | 88426-33-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H26O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 33.33 mg/mL (102.10 mM; Need ultrasonic) ∣DMF : 25 mg/mL (76.59 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (6.13 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1C(CC2CCC(C(C)(C)C)CC2)=C(O)C(C3=C1C=CC=C3)=O |
| Molecular Weight (g/mol) | 326.43 |
| Formula Weight | 326.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction