missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide Beta-D-Glucuronide, TRC

Product Code. 30340690
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30340690 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340690 Supplier Toronto Research Chemicals Supplier No. TRCB6896051MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bumetanide beta-D-Glucuronide
CAS 102623-14-3
Molecular Formula C23H28N2O11S
Quantity 1 mg
Synonym 1-[3-(Aminosulfonyl)-5-(butylamino)-4-phenoxybenzoate]-beta-D-glucopyranuronic Acid, Bumetanide Glucuronide;
InChI Formula InChI=1S/C23H28N2O11S/c1-2-3-9-25-14-10-12(11-15(37(24,32)33)19(14)34-13-7-5-4-6-8-13)22(31)36-23-18(28)16(26)17(27)20(35-23)21(29)30/h4-8,10-11,16-18,20,23,25-28H,2-3,9H2,1H3,(H,29,30)(H2,24,32,33)/t16-,17-,18+,20-,23-/m0/s1
SMILES CCCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-6-[3-(butylamino)-4-phenoxy-5-sulfamoylbenzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 540.54
Formula Weight 540.1414 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.