missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bumetanide Beta-D-Glucuronide, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Bumetanide beta-D-Glucuronide |
| CAS | 102623-14-3 |
| Molecular Formula | C23H28N2O11S |
| Quantity | 10 mg |
| Synonym | 1-[3-(Aminosulfonyl)-5-(butylamino)-4-phenoxybenzoate]-beta-D-glucopyranuronic Acid, Bumetanide Glucuronide |
| InChI Formula | InChI=1S/C23H28N2O11S/c1-2-3-9-25-14-10-12(11-15(37(24,32)33)19(14)34-13-7-5-4-6-8-13)22(31)36-23-18(28)16(26)17(27)20(35-23)21(29)30/h4-8,10-11,16-18,20,23,25-28H,2-3,9H2,1H3,(H,29,30)(H2,24,32,33)/t16-,17-,18+,20-,23-/m0/s1 |
| SMILES | CCCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[3-(butylamino)-4-phenoxy-5-sulfamoylbenzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 540.54 |
| Formula Weight | 540.1414 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction