missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide, MP Biomedicals™

Product Code. 11464601 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11464601

Brand: MP Biomedicals 0215426301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bumetanide is used to probe the role of prostaglandin E2 in the regulation of chloride secretion across porcine endometrial epithelial cells.

  • A study of the effects of bumetanide application and subsequent sodium potassium chloride cotransporter (NKCC1) inhibition in cultured liver cells has indicated increased alpha-1-smooth muscle actin expression in 2-day-cultured hepatic stellate cells.
  • An investigation of lactacidosis treatment of suspended G6 glial cells with bumetanide included at 0.1 mM has shown diminished cell swelling due to lactacidosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Synonym bumetanidum

Specifications

Quantity 1 g
Melting Point 230 to 231°C
Physical Form Powder
Purity ≥99%
Molecular Weight (g/mol) 364.416
Chemical Name or Material Bumetanide

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.