missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bumetanide
Description
Bumetanide is a loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter. More potent than furosemide (Cat. No. 3109). Bumetanide improves electrophysiological, pathological and cognitive deficits in APOE4 knock-in mice. RNA sequencing of neurons derived from APOE4 induced pluripotent stem cell (iPSC) shows Bumetanide reverses Alzheimer's disease transcriptomic signatures.
Specifications
Specifications
| CAS | 28395-03-1 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C17H20N2O5S |
| Formula Weight | 364.42 |
| Purity | 98% |
| Molecular Weight (g/mol) | 364.416 |
| Chemical Name or Material | Bumetanide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?