missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Halogen Bulb, 50w

Product Code. 12922041
Change view
Click to view available options
Light Source:
Halogen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Light Source
12922041 Halogen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12922041 Supplier Fisherbrand™ Supplier No. 3160.6444
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Illuminate Triton II Trinocular Microscopes with this light bulb. Fisherbrand™ Halogen Bulbs are ergonomically designed for everyday use.

  • Halogen bulb, 50w
  • Accessory for microscope: FB70408

Specifications

Light Source Halogen
Voltage 12 V
Wattage 50 W
For Use With (Equipment) Triton II Trinocular Microscopes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.