missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buformin Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Buformin Hydrochloride |
| CAS | 1190-53-0 |
| Analyte or Component Names | Buformin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H15 N5 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Biguanide, 1-butyl-, monohydrochloride, Imidodicarbonimidic diamide, N-butyl-, monohydrochloride, 1-Butylbiguanide hydrochloride, 1-Butylbiguanide, monohydrochloride, 1-Butyldiguanide hydrochloride, Andere, Biforon, Bigunal, Bufonamin, Buformin hydrochloride, Bulbonin, Diabrin, Dibetos, Glibutide, Gliporal, Glybutide, Insulamin, Krebon, N-Butylbiguanide hydrochloride, N1-Butyl-biguanide hydrochloride, N1-n-Butylbiguanide hydrochloride, Panformin, Sindiatil, Tidemol, Ziavetine |
| InChI Formula | InChI=1S/C6H15N5.ClH/c1-2-3-4-10-6(9)11-5(7)8;/h2-4H2,1H3,(H6,7,8,9,10,11);1H |
| Solution Type | Neat |
| SMILES | Cl.CCCCNC(=N)NC(=N)N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction